Orders LinkContactUsLinkComponent
Base materials
Base materials
Yeast extract, Servabacter, 5 kg
Katalog # SERA24540.03
Marketsource
Leverandør:   Serva
Base materials
Katalog # SERA24540.03
Leverandør:  Serva
Marketsource
CAS nummer:  
Base materials
Yeast extract, Servabacter, 5 kg
CAS nummer:  
Price
2.501,66 kr
Log ind for tilgængelighed
1 * 5 kg
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
  • Suitable as nutrient for laboratory media and industrial fermentation
Specifikationer
Specifikationer
  • Pakn:
    5 kg
  • Beskrivelse:
    Yeast extract, Servabacter
Specifikationer