Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Hæmatoxylin

Hæmatoxylin

Glass bottle for solids, 100 g
Katalog # SERA24420.02
Marketsource
Leverandør:   Serva
Hæmatoxylin
Katalog # SERA24420.02
Leverandør:  Serva
Marketsource
CAS nummer:  517-28-2
Hæmatoxylin

Glass bottle for solids, 100 g

CAS nummer:  517-28-2
Price
2.195,48 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  DcH-48, Hydroxybrazilin, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol, NSC 2785, Natural Black I, Hydroxybrasilin, (+)-Hematoxylin
  • CAS nummer:517-28-2
  • Formel:C₁₆H₁₄O₆
  • MDL nummer:MFCD00078111
  • Molecular Weight:302.28 g/mol
  • Merck Index:13,04654
  • Smeltende Pt:291…293 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-237-3
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.