Guanidine thiocyanate
Glass bottle for solids, 1 kg
Katalog # SERA24220.03
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Guanidinium sulphocyanide, GTC, Guanidium thiocyanate, Guanidinium rhodanide
- CAS nummer:593-84-0
- Formel:CH₅N₃·CHNS
- MDL nummer:MFCD00013027
- Molecular Weight:118.16 g/mol
- Smeltende Pt:118 °C
- Tæthed:1,29 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:209-812-1
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg



