Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Glycin

Glass bottle for solids, 1 kg
Katalog # SERA23390.04
Marketsource
Leverandør:   Serva
Glycin
Katalog # SERA23390.04
Leverandør:  Serva
Marketsource
CAS nummer:  56-40-6
Glycin
Glycin

Glass bottle for solids, 1 kg

CAS nummer:  56-40-6
Price
321,11 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  Aminoacetic acid, Glycocoll, H-Gly-OH
  • CAS nummer:56-40-6
  • Formel:NH₂CH₂COOH
  • MDL nummer:MFCD00008131
  • Molecular Weight:75.07 g/mol
  • Merck Index:13,04504
  • Kogende Pt:181 °C (1013 hPa)
  • Smeltende Pt:83 °C
  • Tæthed:1,64 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-272-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.