Glycin
5 kg
SERA23390.03
Marketsource
: Serva
: Aminoacetic acid, Glycocoll, H-Gly-OH
- :56-40-6
- :NH₂CH₂COOH
- :MFCD00008131
- :75.07 g/mol
- :13,04504
- :181 °C (1013 hPa)
- :83 °C
- :1,64 g/cm³ (20 °C)
- :Stue temperatur
- :200-272-2
- Pakn:5 kg