D-(+)-Glucurono-6,3-lactone
Glass bottle for solids, 250 g
Katalog # SERA22900.03
Marketsource
Leverandør: Serva
D-(+)-Glucurono-6,3-lactone
Katalog # SERA22900.03
Leverandør: Serva
Marketsource
CAS nummer: 32449-92-6
Gå til:
Product Details & Documents
Synonymer: (R)-2-((2S,3R,4S)-3,4-Dihydroxy-5-oxotetrahydrofuran-2-yl)-2-hydroxyacetaldehyde, D-(+)-Glucuronolactone
- CAS nummer:32449-92-6
- Formel:C₆H₈O₆
- MDL nummer:MFCD00135622
- Molecular Weight:176.12 g/mol
- EINECS:251-053-3
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:250 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



