Gelatin
Glass bottle, 500 mL
Katalog # SERA22156.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL
- Cat. No.:SERA22156.02
- CAS-nr:9000-70-8
- Protein/peptide name:Gelatin
- Protein synonyms:Gelatine
- Opbevaring betingelser:Ambient



