Folic acid
Glass bottle for solids, 100 g
Katalog # SERA21700.03
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
- CAS nummer:59-30-3
- Formel:C₁₉H₁₉N₇O₆
- MDL nummer:MFCD00079305
- Molecular Weight:441.41 g/mol
- EINECS:200-419-0
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g



