Folic acid
Glass bottle for solids, 25 g
Katalog # SERA21700.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
- CAS nummer:59-30-3
- Formel:C₁₉H₁₉N₇O₆
- MDL nummer:MFCD00079305
- Molecular Weight:441.41 g/mol
- EINECS:200-419-0
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



