2,4-Dihydroxy-5-fluorpyrimidin
Glass bottle for solids, 5 g
Katalog # SERA21670.04
Marketsource
Leverandør: Serva
2,4-Dihydroxy-5-fluorpyrimidin
Katalog # SERA21670.04
Leverandør: Serva
Marketsource
CAS nummer: 51-21-8
Gå til:
Product Details & Documents
Synonymer: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU
- CAS nummer:51-21-8
- Formel:C₄H₃FN₂O₂
- MDL nummer:MFCD00006018
- Molecular Weight:130.08 g/mol
- Merck Index:13,04208
- Kogende Pt:347 °C (1013 hPa)
- Smeltende Pt:282…283 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-085-6
- FN:2811
- ADR:6.1,III
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g



