Orders LinkContactUsLinkComponent
2,4-Dihydroxy-5-fluorpyrimidin
Glass bottle for solids, 1 g
  SERA21670.02
Marketsource
 :   Serva
2,4-Dihydroxy-5-fluorpyrimidin
  SERA21670.02
 :  Serva
Marketsource
 :  51-21-8
2,4-Dihydroxy-5-fluorpyrimidin
Glass bottle for solids, 1 g
 :  51-21-8
1 * 1 g
 :  Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU
  •  :51-21-8
  •  :C₄H₃FN₂O₂
  •  :MFCD00006018
  •  :130.08 g/mol
  •  :13,04208
  •  :347 °C (1013 hPa)
  •  :282…283 °C
  •  :Stue temperatur
  •  :200-085-6
  •  :2811
  •  :6.1,III
  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g