2,4-Dihydroxy-5-fluorpyrimidin
Glass bottle for solids, 1 g
SERA21670.02
Marketsource
: Serva
: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU
- :51-21-8
- :C₄H₃FN₂O₂
- :MFCD00006018
- :130.08 g/mol
- :13,04208
- :347 °C (1013 hPa)
- :282…283 °C
- :Stue temperatur
- :200-085-6
- :2811
- :6.1,III
- Pakning:Glass bottle for solids
- Pakn:1 g