Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

5,5'-Dithiobis(2-nitrobenzosyre)

Glass bottle for solids, 5 g
Katalog # SERA20735.02
Irriterende
Marketsource
Leverandør:   Serva
5,5'-Dithiobis(2-nitrobenzosyre)
Katalog # SERA20735.02
Leverandør:  Serva
Marketsource
CAS nummer:  69-78-3
5,5'-Dithiobis(2-nitrobenzosyre)

Glass bottle for solids, 5 g

CAS nummer:  69-78-3
Price
522,73 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
  • CAS nummer:69-78-3
  • Formel:C₁₄H₈N₂O₈S₂
  • MDL nummer:MFCD00007140
  • Molecular Weight:396.36 g/mol
  • Smeltende Pt:240…244 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-714-4
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.