Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dimethylsulfoxid

Dimethylsulfoxid

Glass bottle, 1 L
Catalog # SERA20385.02
Marketsource
Supplier:   Serva
Dimethylsulfoxid
Catalog # SERA20385.02
Supplier:  Serva
Marketsource
CAS Number:  67-68-5
Dimethylsulfoxid
Dimethylsulfoxid

Glass bottle, 1 L

CAS Number:  67-68-5
Price
418,19 kr
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
  • CAS Number:67-68-5
  • Formula:(CH₃)₂SO
  • MDL Number:MFCD00002089
  • Molecular Weight:78.14 g/mol
  • Merck Index:13,03285
  • Boiling Pt:189 °C (1013 hPa)
  • Melting Pt:18,5 °C
  • Density:1,101 g/cm³ (20 °C)
  • Flash Pt:87 °C
  • Storage Temperature:Stue temperatur
  • EINECS:200-664-3
Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.