Dimethylsulfoxid
Glass bottle, 250 mL
Katalog # SERA20385.01
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
- CAS nummer:67-68-5
- Formel:(CH₃)₂SO
- MDL nummer:MFCD00002089
- Molecular Weight:78.14 g/mol
- Merck Index:13,03285
- Kogende Pt:189 °C (1013 hPa)
- Smeltende Pt:18,5 °C
- Tæthed:1,101 g/cm³ (20 °C)
- Flash Pt:87 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-664-3
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:250 mL



