Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Dextran, M.W. ~ 100,000

Glass bottle for solids, 500 g
Katalog # SERA18693.02
Marketsource
Leverandør:   Serva
Dextran, M.W. ~ 100,000
Katalog # SERA18693.02
Leverandør:  Serva
Marketsource
CAS nummer:  9004-54-0
Dextran, M.W. ~ 100,000

Glass bottle for solids, 500 g

CAS nummer:  9004-54-0
Price
2.255,22 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
  • CAS nummer:9004-54-0
  • Formel:(C₆H₁₀O₅)n
  • MDL nummer:MFCD00130935
  • EINECS:232-677-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.