L(-)-Cystin
Glass bottle for solids, 1 kg
Katalog # SERA17880.03
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
- CAS nummer:56-89-3
- Formel:HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
- MDL nummer:MFCD00064228
- Molecular Weight:240.3 g/mol
- Merck Index:13,02811
- Kogende Pt:428 °C (1013 hPa)
- Smeltende Pt:261…262 °C
- Tæthed:1,677 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-296-3
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg



