Chymostatin
Glass bottle for solids, 25 mg
SERA17158.02
Marketsource
: Serva
- Pakning:Glass bottle for solids
- Pakn:25 mg
- Cat. No.:SERA17158.02
- CAS-nr:9076-44-2
- Protein/peptide name:Chymostatin
- MDL No.:MFCD00071059
- Opbevaring betingelser:Freezer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



