Chloramphenicol
Glass bottle for solids, 10 g
Katalog # SERA16785.01
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
- CAS nummer:56-75-7
- Formel:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
- MDL nummer:MFCD00078159
- Molecular Weight:323.13 g/mol
- Merck Index:13,02087
- Kogende Pt:645 °C (1013 hPa)
- Smeltende Pt:150 °C
- Tæthed:1,49 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-287-4
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 g



