Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cetrimoniumbromid

Cetrimoniumbromid

Glass bottle for solids, 100 g
Katalog # SERA16530.04
Sundhed
Irriterende
Ætsende
Miljø
Marketsource
Leverandør:   Serva
Cetrimoniumbromid
Katalog # SERA16530.04
Leverandør:  Serva
Marketsource
CAS nummer:  57-09-0
Cetrimoniumbromid

Glass bottle for solids, 100 g

CAS nummer:  57-09-0
Price
224,03 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
  • CAS nummer:57-09-0
  • Formel:CH₃(CH₂)₁₅N(Br)(CH₃)₃
  • MDL nummer:MFCD00011772
  • Molecular Weight:364.45 g/mol
  • Merck Index:13,02034
  • Smeltende Pt:237…243 °C
  • Flash Pt:244 °C (closed cup)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-311-3
  • FN:3077
  • ADR:9,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.