Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cetrimoniumbromid

Cetrimoniumbromid

Glass bottle for solids, 500 g
Katalog # SERA16530.02
Sundhed
Irriterende
Ætsende
Miljø
Marketsource
Leverandør:   Serva
Cetrimoniumbromid
Katalog # SERA16530.02
Leverandør:  Serva
Marketsource
CAS nummer:  57-09-0
Cetrimoniumbromid

Glass bottle for solids, 500 g

CAS nummer:  57-09-0
Price
806,50 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
  • CAS nummer:57-09-0
  • Formel:CH₃(CH₂)₁₅N(Br)(CH₃)₃
  • MDL nummer:MFCD00011772
  • Molecular Weight:364.45 g/mol
  • Merck Index:13,02034
  • Smeltende Pt:237…243 °C
  • Flash Pt:244 °C (closed cup)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-311-3
  • FN:3077
  • ADR:9,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer