L(-)-Carnitine
Glass bottle for solids, 25 g
Katalog # SERA16191.01
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
- CAS nummer:541-15-1
- Formel:C₇H₁₅NO₃
- MDL nummer:MFCD00038747
- Molecular Weight:161.20 g/mol
- EINECS:208-768-0
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g



