Conductivity standards, Certipur® Supelco®
KCl 1 M, 111 000 µS/cm, in 500 ml plastic bottle, Plastic bottle, 500 mL
: Merck
These conductivity sachets are convenient, easy to use and ensure accurate and reliable results both in the field or in the laboratory. The 30 ml sachets always provide a fresh solution preventing contamination by microorganisms, CO₂ or other substances. A conductivity sensor can be simply inserted into the sachet.
- Traceable to PTB and NIST
- Tested against primary reference standards
Certipur conductivity standards are also available in 500 ml packs for the user with higher volume requirements.
- Pakning:Plastic bottle
- Pakn:500 mL
- Beskrivelse:KCl 1 M, 111 000 µS/cm, in 500 ml plastic bottle
- Cat. No.:1.01255.0500
- M-Clarity quality level:MQ100



