Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Conductivity standards, Certipur® Supelco®

Conductivity standards, Certipur® Supelco®

KCl 0,1 M, 12 800 µS/cm, in 500 ml plastic bottle, Plastic bottle, 500 mL
Katalog # 1.01254.0500
Leverandør:   Merck
Conductivity standards, Certipur® Supelco®
Katalog # 1.01254.0500
Leverandør:  Merck
CAS nummer:  
Conductivity standards, Certipur® Supelco®
Conductivity standards, Certipur® Supelco®

KCl 0,1 M, 12 800 µS/cm, in 500 ml plastic bottle, Plastic bottle, 500 mL

CAS nummer:  
Price
1.780,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

These conductivity sachets are convenient, easy to use and ensure accurate and reliable results both in the field or in the laboratory. The 30 ml sachets always provide a fresh solution preventing contamination by microorganisms, CO₂ or other substances. A conductivity sensor can be simply inserted into the sachet.

  • Traceable to PTB and NIST
  • Tested against primary reference standards

Certipur conductivity standards are also available in 500 ml packs for the user with higher volume requirements.

Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    500 mL
  • Beskrivelse:
    KCl 0,1 M, 12 800 µS/cm, in 500 ml plastic bottle
  • Cat. No.:
    1.01254.0500
  • M-Clarity quality level:
    MQ100

Specifikationer