Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
X-Gal

X-Gal

Glass bottle for solids, 1 g
Katalog # SERA15243.03
Marketsource
Leverandør:   Serva
X-Gal
Katalog # SERA15243.03
Leverandør:  Serva
Marketsource
CAS nummer:  7240-90-6
X-Gal

Glass bottle for solids, 1 g

CAS nummer:  7240-90-6
Price
746,76 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
Synonymer:  BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
  • CAS nummer:7240-90-6
  • Formel:C₁₄H₁₅BrClNO₆
  • MDL nummer:MFCD00005666
  • Molecular Weight:408.63 g/mol
  • Smeltende Pt:237…239 °C
  • Opbevaringstemperatur:Fryser
  • EINECS:230-640-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.