Brij® 35
Glass bottle for solids, 1 kg
Katalog # SERA15230.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Brij® L23
- CAS nummer:9002-92-0
- Formel:C₅₈H₁₁₈O₂₄
- MDL nummer:MFCD00080891
- Molecular Weight:1199.56 g/mol
- Opbevaringstemperatur:Stue temperatur
- EINECS:500-002-6
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg