Orders LinkContactUsLinkComponent
Natriumpyruvat
Glass bottle for solids, 100 g
  SERA15220.03
Marketsource
 :   Serva
Natriumpyruvat
  SERA15220.03
 :  Serva
Marketsource
 :  113-24-6
Natriumpyruvat
Glass bottle for solids, 100 g
 :  113-24-6
350,98 kr
1 * 100 g
 :  2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
  •  :113-24-6
  •  :CH₃COCOONa
  •  :MFCD00002586
  •  :110.04 g/mol
  •  :220…230 °C
  •  :1,718 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :204-024-4
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g