Orders LinkContactUsLinkComponent
Benzalkoniumchlorid
Plastic bottle, 10 L
  SERA14520.03
Marketsource
 :   Serva
Benzalkoniumchlorid
  SERA14520.03
 :  Serva
Marketsource
 :  63449-41-2
Benzalkoniumchlorid
Plastic bottle, 10 L
 :  63449-41-2
1 * 10 L
 :  Kvaternære ammoniumforbindelser, benzyl-C8-18-alkyldimethyl-, chlorider
  •  :63449-41-2
  •  :C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
  •  :MFCD00145757
  •  :29…34 °C
  •  :0,98 g/cm³ (20 °C)
  •  :264-151-6
  •  :2923
  •  :8,III
  • Pakning:
    Plastic bottle
  • Pakn:
    10 L