Ammoniumnitrat, Supelco®
Plastic bottle, 5 kg
: Merck
- :6484-52-2
- :NH₄NO₃
- :MFCD00011425
- :80.04 g/mol
- :13,00538
- :230 °C (1013 hPa)
- :169 °C
- :1,73 g/cm³ (20 °C)
- :Stue temperatur
- :229-347-8
- :5.1,III
- :M-Clarity™ quality level = MQ200
- :1942
- Pakning:Plastic bottle
- Pakn:5 kg