Benzalkoniumchlorid
Plastic bottle, 2,5 L
Katalog # SERA14520.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Kvaternære ammoniumforbindelser, benzyl-C8-18-alkyldimethyl-, chlorider
- CAS nummer:63449-41-2
- Formel:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
- MDL nummer:MFCD00145757
- Smeltende Pt:29…34 °C
- Tæthed:0,98 g/cm³ (20 °C)
- EINECS:264-151-6
- FN:2923
- ADR:8,III
Specifikationer
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



