Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ammoniumnitrat, Supelco®

Ammoniumnitrat, Supelco®

Plastic bottle, 1 kg
Katalog # 1.01187.1000
Oxiderende
Irriterende
Leverandør:   Merck
Ammoniumnitrat, Supelco®
Katalog # 1.01187.1000
Leverandør:  Merck
CAS nummer:  6484-52-2
Ammoniumnitrat, Supelco®

Plastic bottle, 1 kg

CAS nummer:  6484-52-2
Price
1.280,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
  • CAS nummer:6484-52-2
  • Formel:NH₄NO₃
  • MDL nummer:MFCD00011425
  • Molecular Weight:80.04 g/mol
  • Merck Index:13,00538
  • Kogende Pt:230 °C (1013 hPa)
  • Smeltende Pt:169 °C
  • Tæthed:1,73 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:229-347-8
  • ADR:5.1,III
  •  :M-Clarity™ quality level = MQ200
  • FN:1942
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.