Cellulose
20 kg
Catalog # SERA14204.03
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Microcrystallinecellulose
- CAS Number:9004-34-6
- Formula:(C₁₂H₂₂O₁₁)n
- MDL Number:MFCD00081512
- Molecular Weight:342.3 g/mol
- Merck Index:13,01977
- Melting Pt:260…270 °C
- Density:1,27…1.6 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:232-674-9
Specifications
Specifications
- Pakn:20 kg
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



