Cellulose
Glass bottle for solids, 1 kg
Katalog # SERA14204.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Microcrystallinecellulose
- CAS nummer:9004-34-6
- Formel:(C₁₂H₂₂O₁₁)n
- MDL nummer:MFCD00081512
- Molecular Weight:342.3 g/mol
- Merck Index:13,01977
- Smeltende Pt:260…270 °C
- Tæthed:1,27…1.6 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:232-674-9
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg
- Cat. No.:SERA14204.02



