L (+)-Aspartinsyre
Glass bottle for solids, 1 kg
Katalog # SERA14180.03
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: (S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D
- CAS nummer:56-84-8
- Formel:C₄H₇NO₄
- MDL nummer:MFCD00002616
- Molecular Weight:133.1 g/mol
- Merck Index:13,00845
- Kogende Pt:311 °C (1013 hPa)
- Smeltende Pt:251 °C
- Tæthed:1,6613 g/cm³ (13 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-291-6
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg



