Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

L (+)-Aspartinsyre

Glass bottle for solids, 250 g
Katalog # SERA14180.02
Marketsource
Leverandør:   Serva
L (+)-Aspartinsyre
Katalog # SERA14180.02
Leverandør:  Serva
Marketsource
CAS nummer:  56-84-8
L (+)-Aspartinsyre

Glass bottle for solids, 250 g

CAS nummer:  56-84-8
Price
395,78 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
Synonymer:  (S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D
  • CAS nummer:56-84-8
  • Formel:C₄H₇NO₄
  • MDL nummer:MFCD00002616
  • Molecular Weight:133.1 g/mol
  • Merck Index:13,00845
  • Kogende Pt:311 °C (1013 hPa)
  • Smeltende Pt:251 °C
  • Tæthed:1,6613 g/cm³ (13 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-291-6
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer