Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L(+)-Arginin

L(+)-Arginin

Glass bottle for solids, 1 kg
Katalog # SERA13909.03
Irriterende
Marketsource
Leverandør:   Serva
L(+)-Arginin
Katalog # SERA13909.03
Leverandør:  Serva
Marketsource
CAS nummer:  74-79-3
L(+)-Arginin

Glass bottle for solids, 1 kg

CAS nummer:  74-79-3
Price
1.112,68 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH
  • CAS nummer:74-79-3
  • Formel:C₆H₁₄N₄O₂
  • MDL nummer:MFCD00002635
  • Molecular Weight:174.2 g/mol
  • Merck Index:13,00785
  • Kogende Pt:368 °C (1013 hPa)
  • Smeltende Pt:238 °C
  • Tæthed:1,3 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-811-1
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.