L(+)-Arginin
Glass bottle for solids, 100 g
SERA13909.02
Marketsource
: Serva
: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH
- :74-79-3
- :C₆H₁₄N₄O₂
- :MFCD00002635
- :174.2 g/mol
- :13,00785
- :368 °C (1013 hPa)
- :238 °C
- :1,3 g/cm³ (20 °C)
- :Stue temperatur
- :200-811-1
- Pakning:Glass bottle for solids
- Pakn:100 g