β-Alanin ≥99%, Millipore®
Plastic bottle, 1 kg
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
- CAS Number:107-95-9
- Formula:C₃H₇NO₂
- MDL Number:MFCD00008200
- Molecular Weight:89.09 g/mol
- Merck Index:13,00204
- Boiling Pt:201 °C (1013 hPa)
- Melting Pt:94 °C
- Density:1,44 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Stue temperatur
- EINECS:203-536-5
Documents
Specifications
Specification Test Results
- Assay:99% min
- Heavy metals (as Pb):0.001% max
- Identity (IR-spectrum):passes test
- TLC-test:passes test
Specifications
- Pakning:Plastic bottle
- Pakn:1 kg



