β-Alanin ≥99%, Millipore®
Plastic bottle, 250 g
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
- CAS Number:107-95-9
- Formula:C₃H₇NO₂
- MDL Number:MFCD00008200
- Molecular Weight:89.09 g/mol
- Merck Index:13,00204
- Boiling Pt:201 °C (1013 hPa)
- Melting Pt:94 °C
- Density:1,44 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Stue temperatur
- EINECS:203-536-5
Documents
Specifications
Specification Test Results
- Assay:99% min
- Heavy metals (as Pb):0.001% max
- Identity (IR-spectrum):passes test
- TLC-test:passes test
Specifications
- Pakning:Plastic bottle
- Pakn:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



