L-Alanin
Glass bottle for solids, 500 g
Katalog # SERA11482.03
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
- CAS nummer:56-41-7
- Formel:C₃H₇NO₂
- MDL nummer:MFCD00064410
- Molecular Weight:89.09 g/mol
- Merck Index:13,00203
- Kogende Pt:189 °C (1013 hPa)
- Smeltende Pt:79 °C
- Tæthed:1,4 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-273-8
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



