L-Alanin
Glass bottle for solids, 25 g
Katalog # SERA11482.01
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
- CAS nummer:56-41-7
- Formel:C₃H₇NO₂
- MDL nummer:MFCD00064410
- Molecular Weight:89.09 g/mol
- Merck Index:13,00203
- Kogende Pt:189 °C (1013 hPa)
- Smeltende Pt:79 °C
- Tæthed:1,4 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-273-8
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g



