Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Agarose

Glass bottle for solids, 250 g
Katalog # SERA11397.04
Marketsource
Leverandør:   Serva
Agarose
Katalog # SERA11397.04
Leverandør:  Serva
Marketsource
CAS nummer:  9012-36-6
Agarose
Agarose

Glass bottle for solids, 250 g

CAS nummer:  9012-36-6
Price
Spørg for pris
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
  • CAS nummer:9012-36-6
  • MDL nummer:MFCD00081294
  • Smeltende Pt:60…90 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:232-731-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.