Orders LinkContactUsLinkComponent
EDTA
Glass bottle for solids, 1 kg
  SERA11278.02
Marketsource
 :   Serva
EDTA
  SERA11278.02
 :  Serva
Marketsource
 :  60-00-4
EDTA
EDTA
Glass bottle for solids, 1 kg
 :  60-00-4
440,59 kr
1 * 1 kg
 :  Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
  •  :60-00-4
  •  :C₁₀H₁₆N₂O₈
  •  :MFCD00003541
  •  :292.25 g/mol
  •  :13,03546
  •  :614 °C (1013 hPa)
  •  :220 °C
  •  :0,86 g/cm³ (20 °C)
  •  :>100 °C
  •  :Stue temperatur
  •  :200-449-4
  •  :01-2119486399-18
  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg