EDTA
Glass bottle for solids, 1 kg
SERA11278.02
Marketsource
: Serva
: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
- :60-00-4
- :C₁₀H₁₆N₂O₈
- :MFCD00003541
- :292.25 g/mol
- :13,03546
- :614 °C (1013 hPa)
- :220 °C
- :0,86 g/cm³ (20 °C)
- :>100 °C
- :Stue temperatur
- :200-449-4
- :01-2119486399-18
- Pakning:Glass bottle for solids
- Pakn:1 kg