Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
EDTA

EDTA

Glass bottle for solids, 100 g
Katalog # SERA11278.01
Irriterende
Marketsource
Leverandør:   Serva
EDTA
Katalog # SERA11278.01
Leverandør:  Serva
Marketsource
CAS nummer:  60-00-4
EDTA
EDTA

Glass bottle for solids, 100 g

CAS nummer:  60-00-4
Price
156,82 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
  • CAS nummer:60-00-4
  • Formel:C₁₀H₁₆N₂O₈
  • MDL nummer:MFCD00003541
  • Molecular Weight:292.25 g/mol
  • Merck Index:13,03546
  • Kogende Pt:614 °C (1013 hPa)
  • Smeltende Pt:220 °C
  • Tæthed:0,86 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-449-4
  • NÅ:01-2119486399-18
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.