Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Acetylthiocholiniodid

Acetylthiocholiniodid

Glass bottle for solids, 1 g
Katalog # SERA10570.01
Giftig
Marketsource
Leverandør:   Serva
Acetylthiocholiniodid
Katalog # SERA10570.01
Leverandør:  Serva
Marketsource
CAS nummer:  1866-15-5
Acetylthiocholiniodid
Acetylthiocholiniodid

Glass bottle for solids, 1 g

CAS nummer:  1866-15-5
Price
224,03 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
Synonymer:  (2-Mercaptoethyl)trimethylammonium iodide acetate, 2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide, S-Acetylthiocholine iodide
  • CAS nummer:1866-15-5
  • Formel:H₃CC(S)O(CH₂)₂N(CH₃)₃I
  • MDL nummer:MFCD00011819
  • Molecular Weight:289.18 g/mol
  • Smeltende Pt:204 °C
  • Opbevaringstemperatur:Køleskab
  • EINECS:217-474-1
  • FN:2811
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.