Testkits, BOD til Spectroquant®, Supelco®
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*3 BOD næringssaltmikstur 1.00688.0001 påkrævet
Documents
Specifikationer
Specifikationer
- Pakn:12 Tests
- Beskrivelse:BOD næringssaltblanding (med allylthiourin) til 12 x 1 l næringssaltopløsning
- Til enhed:Prove, Nova 30/60, Multy
- Område:-
- Cat. No.:1.00688.0001
- M-Clarity quality level:MQ100
Specifikationer
Accessories
Produktinformation
- Katalog #ModelTilgængelighedPrice
- Oxygenreaktionsflaske til 1.14662.0001 (MTC)Oxygenreaktionsflaske til 1.14662.0001 (MTC)1.14663.0001MTCLog ind for tilgængelighed1 * 1 Items - 782,00 kr