Forfiltre af glasfiber, Nalgene®
90 mm
Katalog # 513-2812
Leverandør: Thermo Fisher Scientific
Forfiltre af glasfiber, Nalgene®
Katalog # 513-2812
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Increase filtration speed and throughput of highly viscous or heavily particulated solutions by using these glass fiber prefilter discs in combination with any Nalgene™ MF75 filter units or bottle top filters.
Specifikationer
- Størrelse:90 mm
- Cat. No.:513-2812
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning