Forfiltre af glasfiber, Nalgene®
75 mm
Katalog # NALGDS0281-7500
Leverandør: Thermo Fisher Scientific
Forfiltre af glasfiber, Nalgene®
Katalog # NALGDS0281-7500
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Increase filtration speed and throughput of highly viscous or heavily particulated solutions by using these glass fiber prefilter discs in combination with any Nalgene™ MF75 filter units or bottle top filters.
Specifikationer
- Størrelse:75 mm
- Cat. No.:NALGDS0281-7500
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning