Cavity slides
Cavity slides with epoxy coating, sort, 8, 6 mm
Katalog # 631-0451
Leverandør: Epredia
Gå til:
Product Details & Documents
These autoclavable slides are specially designed for immunofluorescence.
- Adhesive surface and hydrophobic mask
- Hydrophobic epoxy- or PTFE-coating
- (*) with rings or wells
Developped for clinical diagnosis applications.
Bestillingsoplysninger: Available with rings or wells (*).
Documents
Specifikationer
- Ant. brønde:8
- Farve:sort
- Beskrivelse:Cavity slides with epoxy coating
- Seq:008
- Brønd Ø:6 mm