Dækglas, Menzel-Gläser
Cover glasses, 24×32 mm, #1,5
Katalog # 630-2873
Leverandør: Epredia
Jump to:
Product Details & Documents
These cover slips are suitable for in vitro diagnostic (IVD) applications.
- Uniform quality of thickness
- No bubbles or flaws
- Made of pre-washed pure white Hydrolytic Class 1 glass
Documents
Specifications
- Antal:#1,5
- Beskrivelse:Cover glasses
- L×B:24×32 mm
- Seq:18