Temperature stabilisation jacket
Silicone O-ring
Katalog # SCOI285405532
Leverandør: SI Analytics
Gå til:
Product Details & Documents
The temperature of capillary viscometers can be stabilised in this jacket using circulation thermostats from 0 to 180 °C.
- Support plate to facilitate the use
- Quick-action seal to simplify the changing viscometers
- Additional hole in the support plate so that a control thermometer can fit
Suitable for use with Ubbelohde viscometers.
Specifikationer
- Beskrivelse:Silicone O-ring
- Seq:102